Learn More
Medchemexpress LLC TLR7/8-IN-1 | 2205095-75-4 | 99.92% | 532.68 | 50 MG
SDP
Catalog No. 5000350079
Encompass_Preferred
TLR7/8-IN-1 is a crystalline form of a TLR7/TLR8 inhibitor extracted from patent WO2019220390, compound 2b. It is intended for research use only and can be used for the research of autoimmune disease.
  • Crystalline form of a TLR7/TLR8 inhibitor.
  • Extracted from patent WO2019220390, compound 2b.
  • Can be used for the research of autoimmune disease.
  • Available in various solid quantities ranging from 1 mg to 100 mg.
  • High purity of 99.92%.
Catalog No. 50-003-50079 Supplier Medchemexpress LLC Supplier No. HY13932350MG
TLR7/8-IN-1 is a crystalline form of a TLR7/TLR8 inhibitor extracted from patent WO2019220390, compound 2b. It is intended for research use only and can be used for the research of autoimmune disease.
  • Crystalline form of a TLR7/TLR8 inhibitor.
  • Extracted from patent WO2019220390, compound 2b.
  • Can be used for the research of autoimmune disease.
  • Available in various solid quantities ranging from 1 mg to 100 mg.
  • High purity of 99.92%.