Learn More
Medchemexpress LLC HY-103039 5mg Medchemexpress, TLR7 agonist 2 CAS:1642857-69-9 Purity:>98%
SDP
Catalog No. 501968025
Encompass_Preferred
Medchemexpress, HY-103039 5mg TLR7 agonist 2 CAS:1642857-69-9 TLR7 agonist 2 is a potent and selective Toll-like Receptor 7 (TLR7) agonist with a LEC of 0.4 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8025 Supplier Medchemexpress LLC Supplier No. HY1030395MG
Medchemexpress, HY-103039 5mg TLR7 agonist 2 CAS:1642857-69-9 TLR7 agonist 2 is a potent and selective Toll-like Receptor 7 (TLR7) agonist with a LEC of 0.4 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.