Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC TLR7 agonist 3 | 1229024-78-5 | 312.41 g/mol | C18H24N4O | 5 MG
SDP

Catalog No. 5000226726
Encompass_Preferred

TLR7 agonist 3 is a small-molecule agonist of Toll-like receptor 7 intended for research use in immunology and inflammation studies. It has been reported to be conjugatable to targeting ligands to influence myeloid-derived suppressor cells.

  • CAS number: 1229024-78-5.
  • Molecular formula: C18H24N4O; molecular weight: 312.41 g/mol.
  • Available package sizes: 1 mg, 5 mg, 10 mg.
  • Solubility: highly soluble in DMSO; ethanol soluble; in vivo vehicle formulations reported.
  • Target: Toll-like receptor 7 (TLR7); pathway: immunology/inflammation.
  • Storage: powder at -20°C (long term) or 4°C (short term); frozen solutions recommended for longer storage.

Catalog No. 50-002-26726 Supplier Medchemexpress LLC Supplier No. HY1176025MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TLR7 agonist 3 is a small-molecule agonist of Toll-like receptor 7 intended for research use in immunology and inflammation studies. It has been reported to be conjugatable to targeting ligands to influence myeloid-derived suppressor cells.

  • CAS number: 1229024-78-5.
  • Molecular formula: C18H24N4O; molecular weight: 312.41 g/mol.
  • Available package sizes: 1 mg, 5 mg, 10 mg.
  • Solubility: highly soluble in DMSO; ethanol soluble; in vivo vehicle formulations reported.
  • Target: Toll-like receptor 7 (TLR7); pathway: immunology/inflammation.
  • Storage: powder at -20°C (long term) or 4°C (short term); frozen solutions recommended for longer storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.