Learn More
Medchemexpress LLC TLR7 agonist 3 | 1229024-78-5 | 99.9% | 312.41 g/mol | C18H24N4O | 100 MG
SDP
Catalog No. 5000226966
Encompass_Preferred
TLR7 agonist 3 is a small-molecule agonist of Toll-like receptor 7 used as a research reagent to stimulate innate immune responses and evaluate anticancer immunomodulatory strategies. It is supplied as a high-purity solid suitable for in vitro and in vivo studies.
  • Potent agonist of Toll-like receptor 7 (TLR7) for immune activation studies.
  • Applicable for conjugation to targeting ligands to direct activity to specific cell types.
  • High purity solid suitable for analytical and biological assays.
  • Soluble in DMSO (100 mg/mL) and ethanol (≥50 mg/mL); compatible with common in vivo formulations at ≥2.5 mg/mL.
  • Offered in multiple pack sizes including 100 mg for dosing flexibility.
Catalog No. 50-002-26966 Supplier Medchemexpress LLC Supplier No. HY117602100MG
TLR7 agonist 3 is a small-molecule agonist of Toll-like receptor 7 used as a research reagent to stimulate innate immune responses and evaluate anticancer immunomodulatory strategies. It is supplied as a high-purity solid suitable for in vitro and in vivo studies.
  • Potent agonist of Toll-like receptor 7 (TLR7) for immune activation studies.
  • Applicable for conjugation to targeting ligands to direct activity to specific cell types.
  • High purity solid suitable for analytical and biological assays.
  • Soluble in DMSO (100 mg/mL) and ethanol (≥50 mg/mL); compatible with common in vivo formulations at ≥2.5 mg/mL.
  • Offered in multiple pack sizes including 100 mg for dosing flexibility.