Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC TLR7 agonist 3 | 1229024-78-5 | MFCD34179471 | 99.9% | 312.41 | C18H24N4O | 25 MG
SDP

Catalog No. 5000227774
Encompass_Preferred

TLR7 agonist 3 is a small-molecule agonist of Toll-like receptor 7 used in preclinical research to activate innate immune responses and to evaluate anticancer immunotherapy approaches.

  • Acts as an agonist of Toll-like receptor 7 (TLR7) in vitro.
  • Used for preclinical immune activation and anticancer research.
  • Molecular formula C18H24N4O.
  • Molecular weight 312.41.
  • Reported high purity (manufacturer lists 99.90%).
  • Soluble in DMSO (100 mg/mL) and ethanol (≥50 mg/mL).
  • Recommended storage: powder -20°C for long term; in solvent -80°C for extended storage.

Catalog No. 50-002-27774 Supplier Medchemexpress LLC Supplier No. HY11760225MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TLR7 agonist 3 is a small-molecule agonist of Toll-like receptor 7 used in preclinical research to activate innate immune responses and to evaluate anticancer immunotherapy approaches.

  • Acts as an agonist of Toll-like receptor 7 (TLR7) in vitro.
  • Used for preclinical immune activation and anticancer research.
  • Molecular formula C18H24N4O.
  • Molecular weight 312.41.
  • Reported high purity (manufacturer lists 99.90%).
  • Soluble in DMSO (100 mg/mL) and ethanol (≥50 mg/mL).
  • Recommended storage: powder -20°C for long term; in solvent -80°C for extended storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.