Learn More
Medchemexpress LLC TLR7 agonist 3 | 1229024-78-5 | MFCD34179471 | 99.9% | 312.41 g/mol | C18H24N4O | 50 MG
SDP
Catalog No. 5000228335
Encompass_Preferred
TLR7 agonist 3 is a synthetic small-molecule agonist of toll-like receptor 7 supplied as a high-purity white to off-white powder for preclinical and research use. It is suitable for biochemical and cellular assays probing innate immune signaling and is available as a solid and as a 10 mM solution in DMSO. Follow recommended storage and handling instructions for stability. For research use only.
  • High purity suitable for assay work (reported 99.9%).
  • Molecular weight 312.41 g/mol; formula C18H24N4O.
  • Available as powder and 10 mM solution in DMSO.
  • Store powder at 4 °C (short term) or -20 °C (long term); store solutions at -20 °C or -80 °C.
  • Intended for laboratory research use only.
Catalog No. 50-002-28335 Supplier Medchemexpress LLC Supplier No. HY11760250MG
TLR7 agonist 3 is a synthetic small-molecule agonist of toll-like receptor 7 supplied as a high-purity white to off-white powder for preclinical and research use. It is suitable for biochemical and cellular assays probing innate immune signaling and is available as a solid and as a 10 mM solution in DMSO. Follow recommended storage and handling instructions for stability. For research use only.
  • High purity suitable for assay work (reported 99.9%).
  • Molecular weight 312.41 g/mol; formula C18H24N4O.
  • Available as powder and 10 mM solution in DMSO.
  • Store powder at 4 °C (short term) or -20 °C (long term); store solutions at -20 °C or -80 °C.
  • Intended for laboratory research use only.