Learn More
Medchemexpress LLC Tlr7 agonist 4 | 2413016-42-7 | 98.9% | C23H34N6O3 | 10MG
SDP
Catalog No. 5000259160
Encompass_Preferred
TLR7 agonist 4 is a small-molecule Toll-like receptor 7 (TLR7) agonist reported with an EC50 of 4.3 nM. It is supplied as a high-purity research reagent (formula C23H34N6O3, MW 442.55) intended for laboratory research use only; not for human or clinical applications.
  • Potent TLR7 agonist with EC50 4.3 nM.
  • High chemical purity (≈98.9%).
  • Well-characterized small molecule for immunology assays.
  • Solid reagent with recommended cold storage for stability.
  • Includes safety data sheet for handling and hazard information.
Catalog No. 50-002-59160 Supplier Medchemexpress LLC Supplier No. HY14596110MG
TLR7 agonist 4 is a small-molecule Toll-like receptor 7 (TLR7) agonist reported with an EC50 of 4.3 nM. It is supplied as a high-purity research reagent (formula C23H34N6O3, MW 442.55) intended for laboratory research use only; not for human or clinical applications.
  • Potent TLR7 agonist with EC50 4.3 nM.
  • High chemical purity (≈98.9%).
  • Well-characterized small molecule for immunology assays.
  • Solid reagent with recommended cold storage for stability.
  • Includes safety data sheet for handling and hazard information.