Learn More
Enzo Life Sciences TLR7 monoclonal antibody (4F4) (100µl)
SDP
Supplier:  Enzo Life Sciences ENZABS7430100
Encompass_Preferred
Clone: 4F4. Isotype: IgG1κ. Immunogen: Recombinant human TLR7 (aa 451-500) purified from E. coli. Source: Purified from ascites. Species reactivity: Human. Applications: ELISA, Flow Cytometry, ICC, IF. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.4, containing 0.02% sodium azide and 10% glycerol.
Catalog No. 50-201-3284