Learn More
Enzo Life Sciences Toll-like receptor 7 polyclonal antibody (100µg)
SDP
Supplier:  Enzo Life Sciences ALX210874C100
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to aa 706-728 of human TLR7 (Toll-like receptor 7). Species reactivity: Human, Mouse, Rat. Applications: Flow Cytometry, ICC, IHC (FS), IHC (PS), IP, WB. Recommended Dilutions: Flow Cytometry (intracellular, 0.5-5µg/106 cells) Immunohistochemistry (paraffin sections, 5-10µg/ml) Western Blot (1-3µg/ml). Purity: Protein G-affinity purified. Formulation: Liquid. In PBS containing 0.05% BSA and 0.05% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-8176