Learn More
Aviva Systems Biology Corp TLR8 Antibody - C-terminal region (ARP59048_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59048_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TLR8. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human TLR8. Alternate names: CD288, MGC119599, MGC119600ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-286-567