Learn More
Enzo Life Sciences Toll-like receptor 8 monoclonal antibody (44C143) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804376C100
Encompass_Preferred
Clone: 44C143. Isotype: IgG1. Immunogen: Synthetic peptide corresponding to aa 750-850 of the cytoplasmic portion of human TLR8 (Toll-like receptor 8). Species reactivity: Human, Mouse. Applications: Flow Cytometry, IHC (PS), WB. Purity: Protein G-affinity purified. Formulation: Liquid. In PBS containing 0.05% BSA and 0.05% sodium azide.
Catalog No. 50-200-9896