Learn More
Aviva Systems Biology Corp TLX1 Antibody - N-terminal region (ARP38630_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38630_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TLX1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TLX1. Alternate names: HOX11, MGC163402, TCL3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-56734