Learn More
ABclonal Technology TLX1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A1286120UL
Encompass_Preferred
This gene encodes a nuclear transcription factor that belongs to the NK-linked or NK-like (NKL) subfamily of homeobox genes. The encoded protein is required for normal development of the spleen during embryogenesis. This protein is also involved in specification of neuronal cell fates. Ectopic expression of this gene due to chromosomal translocations is associated with certain T-cell acute lymphoblastic leukemias. Alternate splicing results in multiple transcript variants.
Catalog No. 50-001-15417