Learn More
Aviva Systems Biology Corp TLX2 Antibody - N-terminal region (ARP39234_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39234_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TLX2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human TLX2. Alternate names: Enx, HOX11L1, NCXApplicationWB. Species reactivity: Cow, Guinea Pig, Human, Rabbit, Rat
Catalog No. 50-284-790