Learn More
Rockland Immunochemicals TM Buffer (50 mM Tris Cl, pH 7.5, 10 mM Magnesium Sulfate) - MB-059
SDP
Catalog No. 501059017
Encompass_Preferred
1X TM Buffer (50 mM Tris HCl, 10 mM Magnesium Sulfate at pH 7.5) is suitable for molecular biology assays.
Catalog No. 50-105-9017 Supplier Rockland Immunochemicals Supplier No. MB059
1X TM Buffer (50 mM Tris HCl, 10 mM Magnesium Sulfate at pH 7.5) is suitable for molecular biology assays.