Learn More
Drg International Inc DRG TM-CA 19-9 ELISA RUO, 96 Well Kit
Supplier:  Drg International Inc EIA5069R
Encompass_Preferred
An enzyme immunoassay for the quantitative measurement of Cancer Antigen 19-9 (CA 19-9) in serum or plasma (EDTA or heparin plasma). This product is for Research Use Only. The DRG TM-CA 19-9 ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA) based on the sandwich principle. The microtiter wells are coated with a monoclonal [mouse] antibody directed towards a unique antigenic site of the CA 19-9 molecule. An aliquot sample containing endogenous CA 19-9 is incubated in the coated well with assay buffer. After a washing step a second incubation follows with enzyme conjugate, which is an anti-CA 19-9 antibody conjugated with horseradish peroxidase.
Catalog No. 50-194-7621