Learn More
Drg International Inc DRG TM-CA 72-4 ELISA RUO, 96 Well Kit
Supplier:  Drg International Inc EIA5071R
Encompass_Preferred
An enzyme immunoassay for the quantitative measurement of Cancer Antigen 72-4 (CA 72-4/TAG-72) in serum and plasma. This product is for Research Use Only. The DRG TM-CA 72-4 ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA) based on the sandwich principle. The microtiter wells are coated with a monoclonal mouse antibody (Clone CC49) directed towards a unique antigenic site on a CA 72-4 molecule. An aliquot sample containing endogenous CA 72-4 is incubated in the coated well with enzyme conjugate, which is an anti-CA 72-4 antibody (Clone B72.3) conjugated with horseradish peroxidase.
Catalog No. 50-194-7622