Learn More
Drg International Inc DRG TM-CYFRA 21-1 ELISA RUO, 96 Well Kit
Supplier:  Drg International Inc EIA5070R
Encompass_Preferred
An enzyme immunoassay for the quantitative measurement of CYFRA (Cytokeratin 19 Fragment) 21-1 in serum and plasma (heparin- or citrate plasma). This product is for Research Use Only. The DRG TM-CYFRA 21-1 ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA) based on the sandwich principle. The microtiter wells are coated with a monoclonal [mouse] antibody directed towards a unique antigenic site of the CYFRA21-1 molecule. An aliquot of patient sample containing endogenous CYFRA21-1 is incubated in the coated well with enzyme conjugate, which is an anti- CYFRA21-1 monoclonal antibody conjugated with horseradish peroxidase.
Catalog No. 50-187-1252