Learn More
Grainger BUSSMANN, Fuse, 10A
Catalog No. NC1912289
Encompass_Preferred
The Bussmann 10 Amp 1CL94 Fast-Acting Glass Tube Fuse is a 10 amp, 250 volt AC/DC fast-acting fuse with a glass tube body and nickel-plated brass endcaps. It has a fuse size of 1CL and features a dual-element time-delay design for circuits and applications where transient current surges occur. This fuse provides overload and short circuit protection for electrical and electronic devices. The glass tube allows for visual inspection of a blown fuse.
Catalog No. NC1912289 Supplier Grainger Supplier No. 1CL94
The Bussmann 10 Amp 1CL94 Fast-Acting Glass Tube Fuse is a 10 amp, 250 volt AC/DC fast-acting fuse with a glass tube body and nickel-plated brass endcaps. It has a fuse size of 1CL and features a dual-element time-delay design for circuits and applications where transient current surges occur. This fuse provides overload and short circuit protection for electrical and electronic devices. The glass tube allows for visual inspection of a blown fuse.