Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NETS PRODUCTS GROUP LLC TM-M30 THERMAL RECEIPT PRINTER

Catalog No. NC1481795
Encompass_Preferred

  • Flexible configuration with both top and front paper exit options.
  • Compact design suitable for space-constrained installations.
  • Robust connectivity options: USB, Ethernet, Bluetooth, and NFC.
  • Compatible with iOS, Android, and Windows operating systems.
  • Supports direct printing from web-based applications using ePOS Print technology.
  • Print method: Thermal line printing.
  • Maximum print speed of 200 mm/sec.
  • Print resolution of 203 dpi.
  • Print width of 3 in (79.5 ± 0.5 mm).
  • Built-in data buffer: 4 KB / 45 Bytes.
  • Dimensions: 127 x 127 x 127 mm.
  • Mass: 1200 g
  • Auto cutter: true.
  • Max paper roll diameter: 83 mm.
  • Paper loading: Drop-in.
  • Warranty: 24 months carry-in.

Catalog No. NC1481795 Supplier NETS PRODUCTS GROUP LLC Supplier No. TMM30
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

  • Flexible configuration with both top and front paper exit options.
  • Compact design suitable for space-constrained installations.
  • Robust connectivity options: USB, Ethernet, Bluetooth, and NFC.
  • Compatible with iOS, Android, and Windows operating systems.
  • Supports direct printing from web-based applications using ePOS Print technology.
  • Print method: Thermal line printing.
  • Maximum print speed of 200 mm/sec.
  • Print resolution of 203 dpi.
  • Print width of 3 in (79.5 ± 0.5 mm).
  • Built-in data buffer: 4 KB / 45 Bytes.
  • Dimensions: 127 x 127 x 127 mm.
  • Mass: 1200 g
  • Auto cutter: true.
  • Max paper roll diameter: 83 mm.
  • Paper loading: Drop-in.
  • Warranty: 24 months carry-in.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.