Learn More
Enzo Life Sciences Thrombomodulin monoclonal antibody (Phx-01) (100µl)
SDP
Supplier:  Enzo Life Sciences ENZABS1100100
Encompass_Preferred
Clone: Phx-01. Isotype: IgG1. Species reactivity: Mammalian. Specificity: Detects an epitope within the EGF-5 domain. Applications: ELISA, IHC, WB, FUNC. Purity: Purified. Formulation: Liquid. In PBS containing 0.03% thimerosal.
Catalog No. 50-201-2667