Learn More
Aviva Systems Biology Corp TM2D2 Antibody - middle region (ARP60590_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60590_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TM2D2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human TM2D2. Alternate names: BLP1, MGC125813, MGC125814ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-287-096