Learn More
Bioworld TM4G Basal Salt Mixture, 25 L
Catalog No. 50254833
Encompass_Preferred
TM4G Basal Salt MixtureContains Macronutrients and MicronutrientsSucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 25L or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 2.88 g/LSoluble in water
Catalog No. 50-254-833 Supplier Bioworld Supplier No. 306301382
TM4G Basal Salt MixtureContains Macronutrients and MicronutrientsSucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 25L or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 2.88 g/LSoluble in water