Learn More
Bioworld TM4G Medium, 50 L
Supplier:  Bioworld 306301773
Encompass_Preferred
Specification:Contains Macronutrients, Micronutrients and vitaminsSucrose, Cytokines, and Auxins should be added as needed Provided as 1 unit to make 25L or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 2.99 g/LSoluble in waterMedia Formulation (mg/L):Cobalt chloride, hexahydrate: 0.025Cupric Sulfate, pentahydrate: 0.025Disodium EDTA: 18.65Ferrous sulfate: 13.9Boric acid: 6.2Manganese sulfate, monohydrate: 16.9Molybdic acid, sodium salt, dihydrate: 0.25Zinc sulfate heptahydrate: 9.2Potassium iodide: 0.83Ammonium nitrate: 320Ammonium phosphate, monobasic: 230Ammonium Sulfate: 130Calcium chloride, anhydrous: 113.5Magnesium sulfate, anhydrous: 122.12Potassium nitrate: 1900Glycine: 2.5Folic acid: 0.5D-Biotin: 0.05Myo-Inositol: 100Nicotinic acid: 5Pyridoxine hydrochloride: 0.5Thiamine hydrochloride: 0.5
Catalog No. 50-254-939