Learn More
Aviva Systems Biology Corp TM4SF20 Antibody - middle region (ARP49875_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49875_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TM4SF20. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TM4SF20. Alternate names: FLJ22800, PRO994, TCCE518ApplicationWB. Species reactivity: Dog, Human, Rabbit, Rat
Catalog No. 50-014-62961