Learn More
Aviva Systems Biology Corp TM4SF4 Antibody - N-terminal region (ARP46480_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46480_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TM4SF4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TM4SF4. Alternate names: FLJ31015, ILTMP, il-TMPApplicationWB. Species reactivity: Cow, Guinea Pig, Human
Catalog No. 50-014-59339