Learn More
Medchemexpress LLC HY-100447 5mg Medchemexpress, TM5275 (sodium) CAS:1103926-82-4 Purity:>98%
SDP
Catalog No. 501931837
Encompass_Preferred
Medchemexpress, HY-100447 5mg TM5275 (sodium) CAS:1103926-82-4 TM5275 sodium is a plasminogen activator inhibitor ( PAI-1 ) with an IC 50 of 6.95 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1837 Supplier Medchemexpress LLC Supplier No. HY1004475MG
Medchemexpress, HY-100447 5mg TM5275 (sodium) CAS:1103926-82-4 TM5275 sodium is a plasminogen activator inhibitor ( PAI-1 ) with an IC 50 of 6.95 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.