Learn More
Medchemexpress LLC TM5275 sodium | 1103926-82-4 | MFCD28396414 | 99.0% | 543.97 | C28H27ClN3NaO5 | 1 MG
SDP
Catalog No. 5000214658
Encompass_Preferred
TM5275 sodium is a small-molecule plasminogen activator inhibitor-1 (PAI-1) inhibitor for research use. It shows inhibitory activity against PAI-1 with an IC50 of 6.95 μM and is supplied as the sodium salt in solid form or as DMSO solutions for assay-ready preparation.
  • Inhibits PAI-1 with reported IC50 of 6.95 μM.
  • Sodium salt form for improved solubility.
  • High purity (≈99%) suitable for research applications.
  • Available as solid and as DMSO solutions for convenience.
  • Molecular formula C28H27ClN3NaO5; molecular weight 543.97.
  • Provides CAS number 1103926-82-4 for unambiguous identification.
Catalog No. 50-002-14658 Supplier Medchemexpress LLC Supplier No. HY1004471MG
TM5275 sodium is a small-molecule plasminogen activator inhibitor-1 (PAI-1) inhibitor for research use. It shows inhibitory activity against PAI-1 with an IC50 of 6.95 μM and is supplied as the sodium salt in solid form or as DMSO solutions for assay-ready preparation.
  • Inhibits PAI-1 with reported IC50 of 6.95 μM.
  • Sodium salt form for improved solubility.
  • High purity (≈99%) suitable for research applications.
  • Available as solid and as DMSO solutions for convenience.
  • Molecular formula C28H27ClN3NaO5; molecular weight 543.97.
  • Provides CAS number 1103926-82-4 for unambiguous identification.