Learn More
Selleck Chemical LLC TM5275 Sodium
SDP
Supplier:  Selleck Chemical LLC S67765MG
Encompass_Preferred
TM 5275 sodium is an orally bioavailable plasminogen activator inhibitor-1 (PAI-1) inhibitor with IC50 of 6 95 M
Catalog No. 50-000-02729