Learn More
Aviva Systems Biology Corp TM7SF2 Antibody - N-terminal region (ARP45456_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45456_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TM7SF2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TM7SF2. Alternate names: ANG1, DHCR14A, NET47ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-63059