Learn More
Aviva Systems Biology Corp TM7SF4 Antibody - N-terminal region (ARP49962_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49962_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TM7SF4. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human TM7SF4. Alternate names: DCSTAMP, FIND, MGC138223, MGC138225, TM7SF4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-471