Learn More
Aviva Systems Biology Corp TM9SF1 Antibody - N-terminal region (ARP44683_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44683_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against TM9SF1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human TM9SF1. Alternate names: HMP70, MP70ApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58869