Learn More
Aviva Systems Biology Corp TM9SF1 Antibody - middle region (ARP44685_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44685_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against TM9SF1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human TM9SF1. Alternate names: HMP70, MP70ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-58871