Learn More
Aviva Systems Biology Corp TM9SF1 Antibody - C-terminal region (ARP58962_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58962_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TM9SF1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human TM9SF1. Alternate names: HMP70, MP70ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-542