Learn More
Strem, An Ascensus Company CAS# [75-24-1]. 200g. Trimethylaluminum, elec. gr. (99.999+%-Al) PURATREM. MFCD00008252
SDP
Catalog No. 501979328
Encompass
CAS# 75-24-1. 200g. Trimethylaluminum, elec. gr. (99.999+%-Al) PURATREM. MFCD00008252. Molecular Weight: 72.09. Molecular Formula: (CH3)3Al. Color/form: colorless liq. Strem# 98-1955. www.strem.com/catalog/v3/98-1955/
Catalog No. 50-197-9328 Supplier Strem, An Ascensus Company Supplier No. 981955200G
Add to Cart
Add to Cart
Request a Quote
CAS# 75-24-1. 200g. Trimethylaluminum, elec. gr. (99.999+%-Al) PURATREM. MFCD00008252. Molecular Weight: 72.09. Molecular Formula: (CH3)3Al. Color/form: colorless liq. Strem# 98-1955. www.strem.com/catalog/v3/98-1955/