Learn More
Perkin Elmer US LLC TMA Compression Probe, 1.5 mm Radius Spherical Tip
Catalog No. 502112015
Encompass_Preferred
Penetration and Compression: Narrow tip compression probes are available for penetration measurements. A broad tip compression probe permits the determination of the compressive modulus and additional properties of soft rubber materials. Part of Quartz 3-Point Bending Systems P/N N5390137 and P/N N5390136.
Catalog No. 50-211-2015 Supplier Perkin Elmer US LLC Supplier No. N5190416
Penetration and Compression: Narrow tip compression probes are available for penetration measurements. A broad tip compression probe permits the determination of the compressive modulus and additional properties of soft rubber materials. Part of Quartz 3-Point Bending Systems P/N N5390137 and P/N N5390136.