Learn More
Medchemexpress LLC HY-D0986 1mg Medchemexpress, TMA-DPH CAS:115534-33-3 Purity:>98%
SDP
Catalog No. 501933228
Encompass_Preferred
Medchemexpress, HY-D0986 1mg TMA-DPH CAS:115534-33-3 TMA-DPH is a hydrophobic fluorescent membrane probe. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3228 Supplier Medchemexpress LLC Supplier No. HYD09861MG
Medchemexpress, HY-D0986 1mg TMA-DPH CAS:115534-33-3 TMA-DPH is a hydrophobic fluorescent membrane probe. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.