Learn More
Strem, An Ascensus Company TMA98% in SwagelokCylinder 25g
SDP
Catalog No. 502034284
Encompass
CAS# 75-24-1. 25g. Trimethylaluminum, min. 98%, 93-1360, contained in high-temp 50 ml Swagelok cylinder (96-1071) for CVD/ALD. MFCD00008252. Molecular Weight: 72.09. Molecular Formula: (CH3)3Al. Color/form: colorless liq. Strem# 98-4004. http://www.strem.com/catalog/v/98-4004/
Catalog No. 50-203-4284 Supplier Strem, An Ascensus Company Supplier No. 98400425G
CAS# 75-24-1. 25g. Trimethylaluminum, min. 98%, 93-1360, contained in high-temp 50 ml Swagelok cylinder (96-1071) for CVD/ALD. MFCD00008252. Molecular Weight: 72.09. Molecular Formula: (CH3)3Al. Color/form: colorless liq. Strem# 98-4004. http://www.strem.com/catalog/v/98-4004/