Learn More
Seracare Life Sciences Inc TMB 1-C MEMBRANE HRP SUBSTRATE
Supplier:  Seracare Life Sciences Inc 54200028
Encompass_Preferred
Most sensitive chromogenic substrate for peroxidase detection. It develops a deep blue color at the reaction site in the presence of peroxidase-labeled conjugates. Recommended for membrane assays and dot blots. 1-component format of 3, 3', 5, 5' - Tetramethylbenzidine, no reagent preparation, hence, very convenient.
Catalog No. 50-674-26