Learn More
Surmodics Inc TMB One Component HRP Membrane Substrate, 1L
SDP
Catalog No. NC0874265
Encompass_Preferred
The BioFX™ one component 3,3’,5,5’-tetramethylbenzidine (TMB) substrate is supplied in a ready-to-use solution for membrane applications. The staining pattern provides a dark blue precipitate with low background to increase the signal-to-noise ratios in western blot, dot blots and line assays.
Catalog No. NC0874265 Supplier Surmodics Inc Supplier No. TMBM100001
The BioFX™ one component 3,3’,5,5’-tetramethylbenzidine (TMB) substrate is supplied in a ready-to-use solution for membrane applications. The staining pattern provides a dark blue precipitate with low background to increase the signal-to-noise ratios in western blot, dot blots and line assays.