Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences TMB 3,3'5,5'-Tetramethylbenzidine 1 GR
SDP

Catalog No. 50980348
Encompass_Preferred

C₁₆H₂₀N₂ F.W. 240.34
Assay: >98%
Melting point: 166-169°C

A non-carcinogenic substitute for DAB and DAB-HCl. Used as a sensitive and specific reagent for the detection of blood. Standefer, J.C., Vanderjagt, D. (1977) Assay of Hemoglobin. Clin. Chem. 23, 749, Boss, E.S., et al., (1981). Assay of Peroxidases. J. of Immunoassay. 2,187. and Holland, V.R., et al., (1974) Tetrahedron, 30, 3299.

Catalog No. 50-980-348 Supplier Electron Microscopy Sciences Supplier No. 13070
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

C₁₆H₂₀N₂ F.W. 240.34
Assay: >98%
Melting point: 166-169°C

A non-carcinogenic substitute for DAB and DAB-HCl. Used as a sensitive and specific reagent for the detection of blood. Standefer, J.C., Vanderjagt, D. (1977) Assay of Hemoglobin. Clin. Chem. 23, 749, Boss, E.S., et al., (1981). Assay of Peroxidases. J. of Immunoassay. 2,187. and Holland, V.R., et al., (1974) Tetrahedron, 30, 3299.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.