Learn More
Cayman Chemical TMB 8 (hydrochloride), Purity: ≥98%, CAS: 53464-72-5, 5 mg
SDP
Catalog No. 502964511
Encompass_Preferred
A non-competitive antagonist of nAChRs (IC50s = 390 and 350 nM, respectively, for human muscle-type and and α3β4 subunit-containing ganglionic nAChRs); an inhibitor of intracellular calcium availability in smooth and skeletal muscle; blocks the contractile response in isolated rabbit aortic strip when used at a concentration of 50 µM; inhibits calcium influx and efflux in isolated guinea pig ileum when used at a concentration of 65 µM; used in the study of intracellular calcium dynamics; dose-dependently inhibits PKC activity
Catalog No. 50-296-4511 Supplier Cayman Chemical Supplier No. 239285 MG
A non-competitive antagonist of nAChRs (IC50s = 390 and 350 nM, respectively, for human muscle-type and and α3β4 subunit-containing ganglionic nAChRs); an inhibitor of intracellular calcium availability in smooth and skeletal muscle; blocks the contractile response in isolated rabbit aortic strip when used at a concentration of 50 µM; inhibits calcium influx and efflux in isolated guinea pig ileum when used at a concentration of 65 µM; used in the study of intracellular calcium dynamics; dose-dependently inhibits PKC activity