Learn More
Seracare Life Sciences Inc TMB BLUESTOP SOLUTION
Catalog No. 5067446
Encompass_Preferred
TMB BlueSTOP Solution is formulated to stop color development in microwell assays and maintain the blue color of TMB. Unlike traditional acid-based TMB stopping solutions, the O.D. value of TMB does not change upon addition of TMB BlueSTOP. The stopped substrate is stable for an extended period and can be read at 650 nm. TMB BlueSTOP is ideal for use with KPL's microwell peroxidase substrates, SureBlue and SureBlue Reserve 1-Component TMB, and TMB 2-Component Microwell Peroxidase Substrate. The reagent is supplied as a convenient, ready-to-use solution that is stable for a minimum of one year.
This item is not returnable. View return policy
Catalog No. 50-674-46 Supplier Seracare Life Sciences Inc Supplier No. 51500024
TMB BlueSTOP Solution is formulated to stop color development in microwell assays and maintain the blue color of TMB. Unlike traditional acid-based TMB stopping solutions, the O.D. value of TMB does not change upon addition of TMB BlueSTOP. The stopped substrate is stable for an extended period and can be read at 650 nm. TMB BlueSTOP is ideal for use with KPL's microwell peroxidase substrates, SureBlue and SureBlue Reserve 1-Component TMB, and TMB 2-Component Microwell Peroxidase Substrate. The reagent is supplied as a convenient, ready-to-use solution that is stable for a minimum of one year.