Learn More
Selleck Chemical LLC TMB dihydrochloride
SDP
Supplier:  Selleck Chemical LLC S6822100MG
Encompass_Preferred
TMB dihydrochloride (3 3 5 5 -Tetramethylbenzidine dihydrochloride hydrate TMB-d BM blue Sure Blue TMB) is an excellent colorimetric substrate for detection of horseradish peroxidase (HRP) labeled probes and is used in staining procedures in immunohistochemistry and in enzyme-linked immunosorbent assays (ELISA)
Catalog No. 50-000-02547