Learn More
Apexbio Technology LLC TMB dihydrochloride 64285-73-0 1g
SDP
Catalog No. 5000091843
Encompass_Preferred
TMB dihydrochloride (CAS 64285-73-0) is a water-soluble chromogenic substrate frequently employed in biochemical assays involving horseradish peroxidase (HRP) It is designed to enable enzyme-catalyzed colorimetric detection by generating an oxidized blue-colored product upon reaction with hydrogen peroxide TMB dihydrochloride exerts its functional role through HRP-catalyzed oxidation in the presence of hydrogen peroxide Based on its chemical reactivity it is also utilized as a probe for interactions with reactive oxygen and nitrogen species such as singlet oxygen hypochlorite hydrogen peroxide and nitrogen dioxide Based on these characteristics TMB dihydrochloride holds research potential in enzyme-linked immunosorbent assays (ELISA) immunohistochemical staining oxidative stress analysis and microbial metabolism studies
Catalog No. 50-000-91843 Supplier Apexbio Technology LLC Supplier No. C70611000
TMB dihydrochloride (CAS 64285-73-0) is a water-soluble chromogenic substrate frequently employed in biochemical assays involving horseradish peroxidase (HRP) It is designed to enable enzyme-catalyzed colorimetric detection by generating an oxidized blue-colored product upon reaction with hydrogen peroxide TMB dihydrochloride exerts its functional role through HRP-catalyzed oxidation in the presence of hydrogen peroxide Based on its chemical reactivity it is also utilized as a probe for interactions with reactive oxygen and nitrogen species such as singlet oxygen hypochlorite hydrogen peroxide and nitrogen dioxide Based on these characteristics TMB dihydrochloride holds research potential in enzyme-linked immunosorbent assays (ELISA) immunohistochemical staining oxidative stress analysis and microbial metabolism studies