Learn More
Apexbio Technology LLC TMB dihydrochloride 64285-73-0 500mg
SDP
Catalog No. 5000094590
Encompass_Preferred
TMB dihydrochloride (CAS 64285-73-0) is a water-soluble chromogenic substrate frequently employed in biochemical assays involving horseradish peroxidase (HRP) It is designed to enable enzyme-catalyzed colorimetric detection by generating an oxidized blue-colored product upon reaction with hydrogen peroxide TMB dihydrochloride exerts its functional role through HRP-catalyzed oxidation in the presence of hydrogen peroxide Based on its chemical reactivity it is also utilized as a probe for interactions with reactive oxygen and nitrogen species such as singlet oxygen hypochlorite hydrogen peroxide and nitrogen dioxide Based on these characteristics TMB dihydrochloride holds research potential in enzyme-linked immunosorbent assays (ELISA) immunohistochemical staining oxidative stress analysis and microbial metabolism studies
Catalog No. 50-000-94590 Supplier Apexbio Technology LLC Supplier No. C7061500
Add to Cart
Add to Cart
TMB dihydrochloride (CAS 64285-73-0) is a water-soluble chromogenic substrate frequently employed in biochemical assays involving horseradish peroxidase (HRP) It is designed to enable enzyme-catalyzed colorimetric detection by generating an oxidized blue-colored product upon reaction with hydrogen peroxide TMB dihydrochloride exerts its functional role through HRP-catalyzed oxidation in the presence of hydrogen peroxide Based on its chemical reactivity it is also utilized as a probe for interactions with reactive oxygen and nitrogen species such as singlet oxygen hypochlorite hydrogen peroxide and nitrogen dioxide Based on these characteristics TMB dihydrochloride holds research potential in enzyme-linked immunosorbent assays (ELISA) immunohistochemical staining oxidative stress analysis and microbial metabolism studies