Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam TMB ELISA Substrate (High Sensitivity), 1000 mL

Catalog No. NC1345221
Encompass_Preferred
Change view
Click to view available options
Quantity:
1000 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NC1345221 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NC1345221 Supplier Abcam Supplier No. AB1715231000ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Abcam TMB ELISA Substrate (High Sensitivity).

  • Species Reactivity: Species independent
  • Storage: Store in the dark

Specifications

Content And Storage Store in the dark
Target Species Non-species Specific
For Use With (Application) ELISA
Quantity 1000 mL
Regulatory Status RUO
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.