Learn More
Sigma Aldrich Fine Chemicals Biosciences TMB Enhanced One Component
Catalog No. 502901507
Encompass
TMB is a substrate for horseradish peroxidase. The substrate system will react with sites on the membrane containing peroxidase producing an insoluble permanent dark blue reaction product. To stop the reaction rinse the membrane with reagent quality water. This substrate is not recommended for microwell or immunohistochemical applications.
Catalog No. 50-290-1507 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. T9455100ML
TMB is a substrate for horseradish peroxidase. The substrate system will react with sites on the membrane containing peroxidase producing an insoluble permanent dark blue reaction product. To stop the reaction rinse the membrane with reagent quality water. This substrate is not recommended for microwell or immunohistochemical applications.