Learn More
Surmodics Inc TMB Extended Range HRP Microwell Substrate, 100mL
SDP
Catalog No. NC1997987
Encompass_Preferred
BioFX™ TMB Extended Range HRP Microwell Substrate is a low-kinetic range substrate for ELISA applications.
Catalog No. NC1997987 Supplier Surmodics Inc Supplier No. TMBX010001
BioFX™ TMB Extended Range HRP Microwell Substrate is a low-kinetic range substrate for ELISA applications.