Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences TMB-HCl, Dihydrochloride TMB Tablets
SDP

Supplier:  Electron Microscopy Sciences 13076

Encompass_Preferred

CAS #64285-73-0 C₁₆H₂₀N2·₂HCl F.W. 313.30
Assay: >98% (Cl)For your convenience TMB-HCl is available in 1 mg tablets that are ready to use.

Catalog No. 50-980-351


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.